Learn More
Thermo Scientific™ Aluminum Dish and Cover, Flat Bottom and Straight Sides
Product Code. 11303468
Click to view available options
Diameter (Metric):
51 mm
63 mm
76 mm
204 mm
Height (Metric):
12.5 mm
25 mm
44 mm
51 mm
No. per Pack:
2/Pk.
6/Pk.
Unit Size:
Pack of 2
Pack of 6
Product Code. 11303468
Brand: Thermo Scientific™ 039077.UD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Ambient temperatures
Material Aluminum
Shape Flat bottom and straight sides
No. per Pack 6/Pk.
Height (Metric) 12.5 mm
Diameter (Metric) 51 mm