Learn More
Thermo Scientific Chemicals Aluminum Dish, Flat Bottom & Straight Sides;Dia (mm), 76;Wall Ht (mm), 25
Click to view available options
Diameter (Metric):
51 mm
76 mm
90 mm
126 mm
204 mm
Height (Metric):
12.5 mm
25 mm
51 mm
No. per Pack:
2/Pk.
6/Pk.
Unit Size:
Pack of 2
Pack of 6
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Bottom Shape | Flat |
| Material | Aluminum |
| Shape | Flat Bottom and Straight Sides |
| Type | Dish |
| No. per Pack | 2/Pk. |
| Height (Metric) | 51 mm |
| Diameter (Metric) | 204 mm |