Learn More
Brady™ Aluminum: ISO Safety Sign - Warning: Floor level obstacle
Sign compliant to ISO 7010.
Brand:  Brady™ 827364
Additional Details : Poids : 0.10700kg
 View more versions of this product
Product Code. 15644707
  • 21.95€ / Each
Expédition estimée
04-02-2021
Add to basket
Description
Description
  • Aluminum
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Global
Aluminum
Indoor and Outdoor
1.016 mm
-40°C to 140°C
Black on Yellow
Triangular
1 Ea.
273 mm
315 mm