missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitride Rod, Round, 10mm diameter, Thermo Scientific™

372.00€

Chemical Identifiers

CAS 24304-00-5
Molecular Formula AlN
Molecular Weight (g/mol) 40.99
MDL Number MFCD00003429
InChI Key PIGFYZPCRLYGLF-UHFFFAOYSA-N
Synonym aluminum nitride, aluminium nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminium nitride grade a, aluminium nitride grade b, aluminium nitride grade c
PubChem CID 90455
ChEBI CHEBI:50884
IUPAC Name alumanylidyneamine
SMILES N#[Al]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471847
SDS View Documents
Thermo Scientific Alfa Aesar
042998.DO
150 mm
372.00€
150mm
View alternative products
Save Up to 
»
Estimated Shipment: 14-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

24304-00-5
40.99
PIGFYZPCRLYGLF-UHFFFAOYSA-N
90455
alumanylidyneamine
AlN
MFCD00003429
aluminum nitride, aluminium nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminium nitride grade a, aluminium nitride grade b, aluminium nitride grade c
CHEBI:50884
N#[Al]

Specifications

24304-00-5
3.26
AlN
aluminum nitride, aluminium nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminium nitride grade a, aluminium nitride grade b, aluminium nitride grade c
N#[Al]
40.99
CHEBI:50884
White to pale yellow rod
Odorless
MFCD00003429
PIGFYZPCRLYGLF-UHFFFAOYSA-N
alumanylidyneamine
90455
Aluminum nitride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 246-140-8

missing translation for 'rtecsNumber' : BD1055000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.