Learn More
Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, C Plane, Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | Aluminum Oxide Substrate, C Plane |
| Name Note | Polished One Side |
| Quantity | 1 Ea. |
| Molecular Formula | Al2O3 |
| MDL Number | MFCD00003424 |
| Solubility Information | Soluble in hexane and toluene. Insoluble in water and diethyl ether. |