Learn More
Brady™ Aluminum with Polyester Overlaminate Radioactive Warning Sign
Product Code. 11710284
Click to view available options
Height (Metric):
100 mm
200 mm
315 mm
Unit Size:
Each
Product Code. 11710284
Brand: Brady™ 241455
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
For Use With (Application) Used for indoor and outdoor safety signs
Height (Metric) 100 mm
Max. Temperature (Metric) 140°C
Thickness 0.271 mm
Width (Metric) 100 mm
Shape Triangular
Displayed Text Warning of Radioactive materials or radiation ioniserenden
Product Type Radioactive Warning Sign
Color Black on Yellow