Learn More
Brady™ Aluminum Sign: Water fog applicator - ISO 7010
Click to view available options
Width (Metric):
100 mm
150 mm
200 mm
Height (Metric):
100 mm
150 mm
200 mm
Unit Size:
Each
Description
- Aluminum
Specifications
Specifications
| Certifications/Compliance | ISO 7010 |
| Alternate Part Number | F/F011/NT-ALU-100X100/1-B |
| Material | Aluminum |
| Quantity | 1 Ea. |
| Height (Metric) | 100 mm |
| Thickness | 1.016 mm |
| Width (Metric) | 100 mm |
| Temperature Range (Metric) | -40°C to 140°C |
| Shape | Square |
| Languages | Global |
| Show More |