missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Standard For ICP, 10,000μg/mL in 5% HCl, SPEX CertiPrep™

Product Code. 10264192 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10264192

Brand: SPEX Certiprep PLAL13X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7429-90-5, 7647-01-0, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula Al
MDL Number MFCD00134029
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Al]
IUPAC Name aluminum
Solvent or Matrix Hydrogen Chloride
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
Quantity 500 mL
ChEBI CHEBI:33629
Grade Assurance
Chemical Name or Material Aluminum Standard
Show More Show Less
compliance-icons
Product Identifier
  • 10,000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, aluminium

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.