Learn More
Aalborg™ Needle Valve for Aluminum Flowmeter
Click to view available options
Airflow Rate:
20000 mL/min.
2400 mL/min.
60000 mL/min.
Waterflow Rate:
1200mL/min.
130mL/min.
3500mL/min.
For Use With (Application):
High flow
Low flow
Medium flow
Unit Size:
Each
Description
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of anodized aluminum bushings are made of plated brass or 316 stainless steel.
Specifications
Specifications
| Type | Standard Valve |
| For Use With (Application) | High flow |
| Airflow Rate | 60000 mL/min. |
| Waterflow Rate | 3500mL/min. |
| Max. Operating Temperature (Metric) | 65°C |