Learn More
Brady™ Aluminum Warning Sign with Polyester Overlaminate
Brand: Brady™ 255200
Additional Details : Weight : 0.04900kg
Specifications
ISO 7010 | |
1 Ea. | |
Indoor and outdoor safety signs | |
0.271 mm | |
-40°C to 140°C | |
Warning of Flammable Materials | |
Black on Yellow |
Aluminum, Polyester | |
Aluminum with Polyester Overlaminate | |
200 mm | |
200 mm | |
Triangular | |
Warning Sign |