Learn More
Aluminum wire, 0.05mm (0.002in) dia, hard, 99.99% (metals basis)
Product Code. p-4781782
Click to view available options
Quantity:
1 m
10 m
5 m
Unit Size:
10m
1m
5m
Product Code. 11318618
Brand: Thermo Scientific Alfa Aesar 040676.G1
Request Bulk or Custom Format
Form: Approx. 0.0053g/m; Temper: as drawn | CAS: 7429-90-5 | Al | 26.98 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]
Specifications
Color Silver
Appearance Silver-colored wire
Density 2.7g/cm3
Quantity 1 m
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Wire
Chemical Name or Material Aluminum, Hard