Learn More
BRAND™ Amber USP Measuring Flask
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
20 mL
250 mL
1000 mL
Unit Size:
Each
Description
Specifications
Specifications
| Closure Material | Polypropylene |
| Color | Amber |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass (3.3) |
| Socket Joint | Ground Joint NS 24/29 |
| Class | A |