missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRA-402, Cl-form, ion-exchange resin, Thermo Scientific Chemicals

Catalog Number p-4259095
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
52439-77-7
MFCD00145564
This item is not returnable. View return policy
52439-77-7
MFCD00145564

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY
Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.60, harmonic mean size: 0.600 to 0.750mm,% <0.300mm: <1.0%,%>1.180mm: <5.0%
Packaging Plastic bottle
Volume Total Capacity (Cl):> 1.20 eq/L
Water Retention Capacity (Cl): 49.0 to 60.0%
Color Yellow
Quantity 250 g
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Amberlite™ IRA-402

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'