missing translation for 'onlineSavingsMsg'
Learn More

Ambroxol hydrochloride, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63712.03

 View more versions of this product

Product Code. 15455899

  • ONLINE EXCLUSIVE PRICE
    49.90€
    44.65€ / 1g
    Save 5.25€
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ambroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.

Solubility
Slightly soluble in water (∼5 mg/ml) or ethanol (∼5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.