Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Brand:  Cytiva 10600099
Additional Details : Weight : 0.08200kg
 View more versions of this product
Product Code. 15209874
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Individual sheets.
  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results
Specifications
Specifications
9 cm
25/Pk.
Nitrocellulose, Inert Polyester
8 cm
Sheet
0.2 μm