Learn More
Cytiva Amersham™ Spacers for Vertical Gel Units
Product Code. 10075264 Shop All Cytiva Products
Click to view available options
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
Width (Metric):
0.1 cm
2 cm
Unit Size:
Pack of 2
Product Code. 10075264
Brand: Cytiva SE611921.0
Only null left
Specifications
For Use With (Equipment) SE 400, SE 600 Vertical Gel Electrophoresis Unit
Type Spacer
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Length (Metric) 16 cm
Width (Metric) 2 cm