missing translation for 'onlineSavingsMsg'
Learn More

Amiloride hydrochloride dihydrate, Thermo Scientific™

Sodium channel blocker

62.71€ - 186.00€

Chemical Identifiers

CAS 17440-83-4
Molecular Formula C6H13Cl2N7O3
Molecular Weight (g/mol) 302.116
MDL Number MFCD00058197
InChI Key LTKVFMLMEYCWMK-UHFFFAOYSA-N
Synonym amiloride hydrochloride dihydrate, modamide, nirulid, amiloride hcl, amiloride hcl dihydrate, amilorid hydrochlorid-2-wasser, unii-fzj37245uc, amiloride hydrochloride usan:usp, amiloride dihydrate hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
PubChem CID 68540
ChEBI CHEBI:2640
IUPAC Name 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide;dihydrate;hydrochloride
SMILES C1(=C(N=C(C(=N1)Cl)N)N)C(=O)N=C(N)N.O.O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15414599
SDS View Documents
Thermo Scientific Alfa Aesar
J62168.03
1g
ONLINE EXCLUSIVE PRICE
64.00€
62.71€ / 1g
Save 1.29€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15424599
SDS View Documents
Thermo Scientific Alfa Aesar
J62168.06
5g
ONLINE EXCLUSIVE PRICE
190.00€
186.00€ / 5g
Save 4.00€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 μM. Amiloride • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

Solubility
Soluble in water at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

17440-83-4
302.116
LTKVFMLMEYCWMK-UHFFFAOYSA-N
68540
3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide;dihydrate;hydrochloride
C6H13Cl2N7O3
MFCD00058197
amiloride hydrochloride dihydrate, modamide, nirulid, amiloride hcl, amiloride hcl dihydrate, amilorid hydrochlorid-2-wasser, unii-fzj37245uc, amiloride hydrochloride usan:usp, amiloride dihydrate hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
CHEBI:2640
C1(=C(N=C(C(=N1)Cl)N)N)C(=O)N=C(N)N.O.O.Cl

Specifications

17440-83-4
MFCD00058197
14,406
Soluble in water at 50mg/ml
C1(=C(N=C(C(=N1)Cl)N)N)C(=O)N=C(N)N.O.O.Cl
302.116
CHEBI:2640
Amiloride hydrochloride dihydrate
C6H13Cl2N7O3
UN2811
amiloride hydrochloride dihydrate, modamide, nirulid, amiloride hcl, amiloride hcl dihydrate, amilorid hydrochlorid-2-wasser, unii-fzj37245uc, amiloride hydrochloride usan:usp, amiloride dihydrate hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
LTKVFMLMEYCWMK-UHFFFAOYSA-N
3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide;dihydrate;hydrochloride
68540
266.09
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 217-958-2

missing translation for 'rtecsNumber' : UQ2275500

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.