missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine, 97+%, Thermo Scientific™

A calcium channel antagonist with potent antioxidant activity

Brand:  Thermo Scientific Alfa Aesar J62242.09

Quantity 10g

 View more versions of this product

Product Code. 15404659

  • 569.00€ / 10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, dimethyl formamide, acetone, chloroform, ethyl acetate and methanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

88150-42-9
408.88
HTIQEAQVCYTUBX-UHFFFAOYNA-N
2162
3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
C20H25ClN2O5
MFCD00864687
amlodipine, norvasc, amlodis, amlodipino, amlodipinum, amlocard, coroval, lipinox, amlor, amlodipinum latin
CHEBI:2668
CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC

Specifications

88150-42-9
MFCD00864687
UN2811
amlodipine, norvasc, amlodis, amlodipino, amlodipinum, amlocard, coroval, lipinox, amlor, amlodipinum latin
HTIQEAQVCYTUBX-UHFFFAOYNA-N
3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
2162
408.88
Amlodipine
C20H25ClN2O5
10g
14,491
Soluble in dimethyl sulfoxide,ethanol,water,dimethyl formamide,acetone,chloroform,ethyl acetate and methanol.
CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC
408.88
CHEBI:2668
≥97%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.