Learn More
Hanna Instruments™ Ammonia Portable Photometer Kit
Product Code. 11617130
Click to view available options
Type:
Photometer
Unit Size:
Each
Product Code. 11617130
Brand: Hanna Instruments™ HI96715C
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use in measurement of ammonia (as NH3-N) in freshwater samples. Hanna Instruments™ Ammonia Portable Photometer Kit features an advanced optical system.
  • Combination of light emitting diode (LED), narrow band interference filter, and silicon photodetector ensure accurate photometric readings every time
  • CAL Checktrade; feature utilizes ready-made standards traceable to NIST Standard for performance verification and calibration
  • Exclusive cuvette locking system ensures that cuvette is inserted into measurement cell in same position every time to maintain consistent path length
  • Combines accuracy and ease of use in ergonomic, portable design
Specifications
Type Photometer
Accuracy ±0.05mg/L ±5% of reading
For Use With (Application) Measurement of ammonia (as NH3-N) in freshwater samples
Includes Portable Photometer, Two sample cuvettes, Two Caps, CAL Check standards with certificate, Cuvette cleaning cloth, Rugged carrying case, Thermoformed insert for meter and accessories
Light Source LED
Range 0.00 to 9.99mg/L (ppm) (as NH₃−N)
Resolution 0.01 mg/L
Weight (Metric) 360 g