Learn More
Macherey-Nagel™ Ammonia Test, For Aquariums
Allows for the fast determination of ammonium in solutions to keep aquariums in good condition, so fish stay healthy. Macherey-Nagel™ Ammonia Test, For Aquariums provides safe, reliable results within 40 seconds—just dip, swirl and read.
Brand: Macherey-Nagel™ 90714
Additional Details : Weight : 0.80000kg
Description
p>In aquarium water, ammonium is produced by the bacterial decomposition of uneaten food and plant matter as well as fish waste. Although ammonium is an important nutrient for plants, it can convert to ammonia, which is toxic for the fish. Testing the presence of ammonium provides important information about aquarium water quality. If ammonium is detected, a partial water change must be carried out. If necessary, the water pH level must be lowered to avoid dangerous ammonia build-up.Specifications
Ammonia (NH4+) | |
Test strip | |
0 - 0.5 - 1 - 3 - 6mg/L NH4+ | |
25 | |
0 - >6mg/L | |
Store in cool, dry location (below 30°C), away from light | |
Ammonia test strip |
Bright yellow to Blue | |
Determination of ammonia in aquarium water | |
Box | |
1 Each | |
7 x 60 mm | |
40 sec. |