Learn More
STEMCELL Technologies Ammonium Chloride Solution
Description
- Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
- It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
- It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Specifications
Specifications
| Quantity | 500 mL |
| For Use With (Application) | Lysis of Red Blood Cells |