missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 98+%, for biochemistry, Thermo Scientific Chemicals

Catalog Number 12625117
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
N.OP(O)(O)=O
This item is not returnable. View return policy
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
N.OP(O)(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 190.0°C
Quantity 1 kg
Assay Percent Range 98+%
Linear Formula NH4H2PO4
Merck Index 15,541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Formula Weight 115.03
Percent Purity ≥98%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Ammonium dihydrogen phosphate, for biochemistry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'