missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog Number p-3256882
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
12125-01-8
NH4F
MFCD00011423
This item is not returnable. View return policy
12125-01-8
NH4F
MFCD00011423

It is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98.0%
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04
Grade ACS
Sensitivity Hygroscopic
Density 1.01 g/mL
Chemical Name or Material Ammonium fluoride, For analysis ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'