missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexabromoosmate(IV), 99.9% (metals basis), Os 26.5 % min

Product Code. 11387487
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11387487

Brand: Thermo Scientific Alfa Aesar 012648.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24598-62-7 | Br6H8N2Os | 705.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24598-62-7
Molecular Formula Br6H8N2Os
Molecular Weight (g/mol) 705.73
MDL Number MFCD00049389
InChI Key ZPHFWWAVHILXPL-UHFFFAOYSA-J
Synonym diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
PubChem CID 11136267
IUPAC Name diazanium;hexabromoosmium(2-)
SMILES [NH4+].[NH4+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Os+4]

Specifications

Color Black
Physical Form Powder
Quantity 5 g
Formula Weight 705.73
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Os 26.5 % min.
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexabromoosmate(IV)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.