Learn More
Ammonium hydroxide solution, Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis, Honeywell Fluka™
Puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Brand: Honeywell Fluka 05003-2.5L-GL
Additional Details : CAS : 1336-21-6 Poids : 2.27500kg Transport : UN number : 2672 Chem class : 8 Pack group : III
Packaging | Glass Bottle |
---|---|
Quantity | 2.5L |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1336-21-6 | |
35.046 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
14923 | |
azanium;hydroxide |
H5NO | |
MFCD00066650 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
CHEBI:18219 | |
[NH4+].[OH-] |
Specifications
Ammonium Hydroxide Solution | |
1336-21-6 | |
H5NO | |
0.892 to 0.910g/mL at 20°C, 0.9g/mL at 25°C | |
0.27 | |
3587154 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
azanium;hydroxide | |
14923 | |
35.05g/mol | |
Glass Bottle |
Meets analytical specification of Ph.Eur. | |
25 to 30% NH3 basis | |
NH4OH | |
MFCD00066650 | |
2672 | |
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
[NH4+].[OH-] | |
35.046 | |
CHEBI:18219 | |
puriss. | |
2.5L |
Safety and Handling
- Ammonium hydroxide solutio
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ammonia, aqueous solution