missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(II) Sulfate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
3 kg
Specifications
Melting Point | 100°C |
Color | Green |
pH | 3,5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 392.14g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'