Learn More
Ammonium molybdate tetrahydrate, ACS Reagent, Honeywell Fluka™
ACS Reagent, 81.0-83.0% MoO3O basis
Brand: Honeywell Fluka A7302-2.5KG
Additional Details : CAS : 12054-85-2 Poids : 2.50000kg
Quantity | 2.5kg |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
12054-85-2 | |
MFCD00167059 |
Specifications
Ammonium molybdate tetrahydrate | |
(NH4)6Mo7O24 · 4H2O | |
NONH for all modes of transport | |
ACS Reagent | |
2.498g/mL at 25°C |
12054-85-2 | |
MFCD00167059 | |
1235.86g/mol | |
81 to 83% (MoO3 basis) | |
2.5kg |