missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate, ACS, 95% min, Thermo Scientific™

In fertilizers, making nitrous oxide

50.30€ - 148.00€

Chemical Identifiers

CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.043
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name azanium;nitrate
SMILES [NH4+].[N+](=O)([O-])[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341749
SDS View Documents
Thermo Scientific Alfa Aesar
012363.36
500g
ONLINE EXCLUSIVE PRICE
51.10€
50.30€ / 500g
Save 0.80€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11351749
SDS View Documents
Thermo Scientific Alfa Aesar
012363.A3
2kg
ONLINE EXCLUSIVE PRICE
150.00€
148.00€ / 2kg
Save 2.00€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6484-52-2
80.043
DVARTQFDIMZBAA-UHFFFAOYSA-O
22985
azanium;nitrate
H4N2O3
MFCD00011425
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
CHEBI:63038
[NH4+].[N+](=O)([O-])[O-]

Specifications

6484-52-2
H4N2O3
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
DVARTQFDIMZBAA-UHFFFAOYSA-O
azanium;nitrate
22985
80.04
1.72°C
95% min
MFCD00011425
Highly soluble in water; soluble in alcohol and liquid ammonia.
[NH4+].[N+](=O)([O-])[O-]
80.043
CHEBI:63038
ACS
Ammonium nitrate, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P220-P221-P264b-P271-P280-P280i-P305+P351+P338-P370+P380+P375-P372-P380-P501c

H272-H319

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III

missing translation for 'einecsNumber' : 229-347-8

missing translation for 'rtecsNumber' : BR9050000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.