missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Nitrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072829
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
6484-52-2
H4N2O3
80.04
MFCD00011425
DVARTQFDIMZBAA-UHFFFAOYSA-O
ammonium nitrate
[NH4+].[O-][N+]([O-])=O
Boiling Point 210°C
Melting Point 169°C
Color White
pH 4.5 to 6.0
Physical Form Solid
Quantity 500 g
Formula Weight 80.04g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Ammonium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'