missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate Monohydrate, Honeywell Fluka™
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 250 g |
Linear Formula | (NH4)2C2O4 · H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3759830 |
Formula Weight | 142.11g/mol |
Percent Purity | 99.5 to 101% |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.02% (as SO4) |
Chemical Name or Material | Ammonium Oxalate Monohydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ammonium oxalate-1-hydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'