missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, ≥98.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥98.0%

Brand:  Honeywell Fluka 248614-4X2.5KG

Additional Details : CAS : 7727-54-0 Poids : 10.00000kg Transport : UN number : 1444 Chem class : 5.1 Pack group : III


Packaging Poly Bottle
Quantity 4 x 2.5kg

 View more versions of this product

Product Code. 15662700

  • 688.63€ / Pack of 4


Expédition estimée
12-11-2020
Add to basket

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
diazanium;sulfonatooxy sulfate

Specifications

Ammonium Persulfate
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.2g/mol
ACS Reagent
Poly Bottle
7.9 (vs air)
Colorless
4 x 2.5kg
7727-54-0
(NH4)2S2O8
UN1444
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
≥98.0%
1.98g/cm3
≤0.05%
Solid
120°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium persulfate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents