missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, puriss., Honeywell Fluka™
BIOPHARMA

Catalog Number p-7071714
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
N.N.OP(O)(O)=O
7783-28-0
H9N2O4P
132.06
MFCD00010891
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
24540
CHEBI:63051
diazanium;hydrogen phosphate
N.N.OP(O)(O)=O

Puriss., meets analytical specification of NF, 99-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity 99 to 102%
Grade puriss.
Chemical Name or Material Ammonium Phosphate Dibasic, Meets analytical specification of NF
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'