Learn More
Honeywell Fluka™ Ammonium Phosphate Monobasic, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS reagent,≥99.0% (T)
Brand: Honeywell Fluka™ 09717-250G
Additional Details : CAS : 7722-76-1 Poids : 0.25000kg
Quantity | 250g |
---|
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7722-76-1 | |
115.025 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
24402 | |
azanium;dihydrogen phosphate |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
CHEBI:62982 | |
[NH4+].OP(=O)(O)[O-] |
Specifications
Ammonium Phosphate Monobasic | |
H6NO4P | |
MFCD00003396 | |
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
[NH4+].OP(=O)(O)[O-] | |
115.025 | |
CHEBI:62982 | |
≥99% (T) | |
250g |
7722-76-1 | |
NH4H2PO4 | |
NONH for all modes of transport | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
azanium;dihydrogen phosphate | |
24402 | |
115.03g/mol | |
ACS/puriss. p.a. |