missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, ≥98.0%, ACS Reagent, Honeywell Fluka™

Catalog Number p-7120193
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]
This item is not returnable. View return policy
7773-06-0
H6N2O3S
114.119
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
24482
CHEBI:81950
azanium;sulfamate
[NH4+].NS(=O)(=O)[O-]

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 131°C to 135°C
Color White
Quantity 100 g
Linear Formula NH4SO3NH2
UN Number UN3077
Formula Weight 114.12g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.10%
Appearance Crystalline
Density 1g/cm3
Chemical Name or Material Ammonium Sulfamate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'