missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide Solution, 20% in H2O, Honeywell™ Fluka™

Product Code. 15648480 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
25mL
500mL
This item is not returnable. View return policy

Product Code. 15648480

Brand: Honeywell-Fluka A195225ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Color Yellow
Quantity 25 mL
Linear Formula (NH4)2S
UN Number UN2683
Flash Point 32°C (89.6°F)
Formula Weight 68.14g/mol
Concentration or Composition (by Analyte or Components) 20% in H2O
Odor Stench
Packaging Glass Bottle
Appearance Liquid
Vapor Pressure 600 hPa at 20°C
Chemical Name or Material Ammonium sulfide solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.