missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog Number p-4781794
Click to view available options
Quantity:
1 L
4 L
7732-18-5
NH4SCN
MFCD00011428
7732-18-5
NH4SCN
MFCD00011428

Ammonium thiocyanate, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Color Colorless
Formula Weight 76.12
Density 1g/cm3 at 20°C
Solubility Miscible with water, ethanol, methanol, ammonia and acetone
Physical Form Liquid
Chemical Name or Material Ammonium thiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'