missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxapine-d8, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Amoxapine-d8 |
| CAS | 1189671-27-9 |
| Molecular Formula | C17H8D8ClN3O |
| Quantity | 1 mg |
| Synonym | Amoxapine D8 |
| InChI Formula | InChI=1S/C17H16ClN3O/c18-12-5-6-15-13(11-12)17(21-9-7-19-8-10-21)20-14-3-1-2-4-16(14)22-15/h1-6,11,19H,7-10H2/i7D2,8D2,9D2,10D2 |
| SMILES | [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C2=Nc3ccccc3Oc4ccc(Cl)cc24)C1([2H])[2H] |
| IUPAC Name | 8-chloro-6-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)benzo[b][1,4]benzoxazepine |
| Molecular Weight (g/mol) | 321.83 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction