missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ AMTAX sc Reagent

Product Code. 13276848
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
This item is not returnable. View return policy

Product Code. 13276848

Brand: Hach Lange™ BCF1009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Format Liquid
Quantity 2.5 L
compliance-icons
Product Identifier
  • AMTAX sc, Reagenz/Reagent/Réactif
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide; caustic soda, Ethylenedinitrilotetraaceticacid disodium saltdihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.