Learn More
Agilent BioTek AMX Cassette
GREENER_CHOICE
Product Code. 16572102 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
MFPX2
Unit Size:
Each
Product Code. 16572102
Brand: Agilent BioTek 1260035
Two cassettes are required for use with the AMX module (PN 1260041). User installs cassettes with AMX manifolds (included with AMX module). For use with MFXP2R, order two cassettes (does not require PN 1260041).
Dispensing speeds:
  • 96 wells, 5 μL cass, 10 μL/ well: 3 seconds
  • 96 wells, 5 μL cass, 20 μL/ well: 3.5 seconds
  • 384 wells, 5 μL cass, 5 μL/ well: 6.5 seconds
  • 384 wells, 10 μL cass, 10 μL/ well: 8 seconds
  • 384 wells, 1 μL cass, 1 μL/ well: 6 seconds
  • 1536 wells, 1 μL cass, 1 μL/ well: 21 seconds
TRUSTED_SUSTAINABILITY
Specifications
Accuracy <5%
Autoclavable Autoclavable
For Use With (Application) 2 D and 3 D Cell Culture to Concentration Normalization Assays, ELISA, Bead-Based Assays and More
For Use With (Equipment) MFPX2
Includes Two 8-channel autoclavable manifolds, tube-in-tube design
Precision <5% CV
Electrical Requirements 50/60 Hz