Learn More
Epredia™ Lab Vision™ Amyloid A Component Ab-1 Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.
134.64€ - 505.00€
Specifications
Antigen | Amyloid A Component Ab-1 |
---|---|
Clone | mcl |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12679316
|
Epredia™
MS-1219-S1 |
Concentrated | 500 μL |
434.00€
500µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649316
|
Epredia™
MS-1219-S |
Concentrated | 1 mL |
505.00€
1mL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12669316
|
Epredia™
MS-1219-S0 |
Concentrated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12659316
|
Epredia™
MS-1219-R7 |
Prediluted | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Host Species: Mouse
Clone: mcl
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Immunogen: Human amyloid A
Positive Control: Amyloid deposits in kidney
Cellular Localization: Extracellular
Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Spécification
Amyloid A Component Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
mcl | |
Monoclonal | |
Mouse | |
Neuroscience | |
Human amyloid A | |
Primary |