missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMZ 30

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AMZ 30 is a potent, selective and irreversible inhibitor of protein phosphatase methylesterase-1 (PME-1; IC50 = 600 nM) with > 100-fold selectivity for PME-1 over other serine hydrolases in human cell lysates. AMZ 30 inactivates PME-1 and reduces the demethylated form of serine/threonine protein phosphatase 2A (PP2A) in HEK 293T cells. AMZ 30 perturbs leucine carboxyl methyltransferase-1 (LCMT1)-PME-1 methylation equilibrium leading to shortening of mitotic spindles and mitotic arrest in HeLa cells. It also shows antiproliferative properties in an in vitro model of endometrial carcinoma.
Specifications
Specifications
| Chemical Name or Material | (2 E)-2-[(4-Fluorophenyl)sulfonyl]-3-[1-[(3-nitrophenyl)sulfonyl]-1 H-pyrrol-2-yl]-2-propenenitrile |
| CAS | 1313613-09-0 |
| Quantity | 10 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C19H12FN3O6S2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction