missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Anabasine, tech. 85%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar L12089.03

Quantity 1g

Product Code. 15448949

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

13078-04-1
270°C to 272°C
C10H14N2
MFCD00006370
UN3140
14,619
MTXSIJUGVMTTMU-UHFFFAOYNA-N
3-(piperidin-2-yl)pyridine
2181
162.24
Technical
1.046
93°C (199°F)
1.544
1g
82639
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
C1CCC(NC1)C1=CC=CN=C1
162.24
CHEBI:28986
85%
(+/-)-Anabasine
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.