Learn More
Abeba™ Anatom 1122 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 1122/38
Additional Details : Weight : 1.18000kg
Description
- CE EN ISO 20345:2011 S1 SRC
- Velours with breathable textile inlays
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S1 SRC | |
Velour | |
Slip-resistant | |
1122 |
Black | |
38 | |
Polyurethane | |
Steel |