Learn More
Abeba™ Anatom 1122 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 1122/38
Additional Details : Weight : 1.18000kg
 View more versions of this product
Product Code. 15686967
  • 109.42€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Velours with breathable textile inlays
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Velour
Slip-resistant
1122
Black
38
Polyurethane
Steel