Learn More
Abeba™ Anatom 2136 Shoes
Product Code. 15617867
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15617867 36 1 pair
15627867 37 1 pair
15637867 38 1 pair
15647867 39 1 pair
15657867 40 1 pair
15667867 41 1 pair
15677867 42 1 pair
15687867 43 1 pair
15697867 44 1 pair
15607877 45 1 pair
15617877 46 1 pair
15627877 47 1 pair
15637877 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15617867 Supplier Abeba™ Supplier No. 2136/36
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color Black
Material Microfiber
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2136
Toe Material Steel