Learn More
Abeba™ Anatom 2136 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 2136/40
Additional Details : Weight : 1.15000kg
 View more versions of this product
Product Code. 15657867
  • 109.77€ / 1 pair
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
2136
Black
40
Polyurethane
Steel