Learn More
Abeba™ Anatom 2168 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 2168/39
Additional Details : Weight : 1.34000kg
Description
- CE EN ISO 20345:2011 S2 SRC
- Smooth leather
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Scratch-resistant coating in heel and toe area
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Leather | |
Slip-resistant | |
2168 |
Black | |
39 | |
Polyurethane | |
Steel |