Learn More
Abeba™ Anatom 2177 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 2177/39
Additional Details : Weight : 1.28000kg
 View more versions of this product
Product Code. 15617987
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Highly abrasion resistant breathable multifunctional upper material run-dry
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
  • Scratch-resistant coating in heel area
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
39
Polyurethane
Steel
Black
Slip-resistant
2177