Learn More
Abeba™ Anatom 2177 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 2177/42
Additional Details : Weight : 1.28000kg
Description
- CE EN ISO 20345:2011 S2 SRC
- Highly abrasion resistant breathable multifunctional upper material run-dry
- Black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
- Discontinued model
- Scratch-resistant coating in heel area
Specifications
CE EN ISO 20345:2011 S2 SRC | |
42 | |
Polyurethane | |
Steel |
Black | |
Slip-resistant | |
2177 |