Learn More
Abeba™ Anatom 2178 Shoes
Product Code. 15647997
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15647997
Brand: Abeba™ 2178/39
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S2 SRC
  • Highly abrasion resistant breathable multifunctional upper material run-dry
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Discontinued model
  • Scratch-resistant coating in heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color Black
Size 39
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2178
Toe Material Steel